missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Avermectin, ACROS Organics™

Catalog No. AC468392500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468392500 250 mg
AC468390010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC468392500 Supplier Thermo Scientific™ Supplier No. 468392500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Name or Material Avermectin
CAS 65195-55-3
Quantity 250 mg
Molecular Formula C48 H72 O14
Formula Weight 873.09
Physical Form Crystalline Powder
Color White to Light-yellow