Learn More
Cytiva AutoSeq G-50 Columns
Use for rapid removal of unincorporated fluorescent dye-terminators from cycle sequencing reactions prior to automated sequencing. Cytiva Life Sciences™ illustra™ AutoSeq G-50 Columns provide effective purification essential for high-quality sequencing results.
$321.08 - $1440.44
Specifications
For Use With (Application) Rapid removal of unincorporated Fluorescent dye-terminators from cycle sequencing reactions prior to automated sequencing
Product Type Column
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
45-001-400
View Documents
Cytiva
27534001
50 Columns
Each for $321.08
Only null left
45-001-401
View Documents
Cytiva
27534002
250 Columns
Each for $1,440.44
Only null left
Description
Description
  • Columns are prepacked with Sephadex G-50 DNA Grade F and pre-equilibrated in double-distilled water with 0.05% Kathon CG/ICP Biocide added as a preservative
  • Columns that are ready to use enable sample application to collection of purified product in less than 4 min.
  • Can be used to clean up reactions prepared with the BigDye Terminator v3.1 Cycle Sequencing Kit from Applied Biosystems
  • Typical sample volume 12 to 25 μl
  • Suitable for purification of DNA greater than 20 bases in length
  • Optimized for automated fluorescent sequencing
Specifications
Specifications
Rapid removal of unincorporated Fluorescent dye-terminators from cycle sequencing reactions prior to automated sequencing
Column
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'