missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations

Increase pipetting performance with these automation tips, which undergo a rigorous 15-point quality control program to ensure accuracy and consistency.

$128.67 - $1447.47

Specifications

Color Clear
Format Rack
Tip Style Automation
For Use With (Equipment) Zymark/Caliper Liquid Handling Workstations
Product Line Caliper Zymark
View More Specs

Products 7
Catalog Number Mfr. No. Volume (Metric) Filtered Sterility Fit for Purpose Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Filtered Sterility Fit for Purpose Price Quantity  
10-371-033
View Documents
Thermo Scientific™
920021
50 μL Filtered Sterile Sterile and PCR Ready
Case of 4800 Each for $1,447.47
Only null left
 
10-371-032
View Documents
Thermo Scientific™
MB920261
50 μL Non-filtered Sterile Sterile
Case of 4800 Each for $877.27
Only null left
 
21-403-444
View Documents
Thermo Scientific™
920262
50 μL Non-filtered Non-sterile Certified Quality
Pack of 960 Each for $138.03
Case of 5 Pack for $690.15
Only null left
 
21-402-305
View Documents
Thermo Scientific™
921262
100 μL Non-filtered Non-sterile Certified Quality
Pack of 960 Each for $138.03
Case of 5 Pack for $690.15
Only null left
 
21-402-300
View Documents
Thermo Scientific™
923021
150 μL Filtered Sterile Sterile and PCR Ready
Pack of 960 Each for $235.70
Case of 5 Pack for $1,178.50
Only null left
 
10-371-034
View Documents
Thermo Scientific™
MB923261
200 μL Non-filtered Sterile Sterile
Case of 4800 Each for $716.43
Only null left
 
21-402-302
View Documents
Thermo Scientific™
923262
200 μL Non-filtered Non-sterile Certified Quality
Pack of 960 Each for $128.67
Case of 5 Pack for $643.35
Only null left
 
Description

Description

Increase pipetting performance with Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Caliper/Zymark RapidPlate, Caliper Presto, Allegro, Zephyr and SciClone.

Features:
  • Available as sterile and filtered.
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
  • Lot-tested during production on actual workstation for which they are created.
  • Low percentage coefficient of variation (%CV).
Warranty:A five (5)-year shelf life from the date of manufacture is recommended.
Specifications

Specifications

Clear
Automation
Caliper Zymark
Rack
Zymark/Caliper Liquid Handling Workstations
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'