missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations

Catalog No. 10371034
Change view
Click to view available options
Volume (Metric):
50 μL
100 μL
150 μL
200 μL
Filtered:
Filtered
Non-filtered
Sterility:
Non-sterile
Sterile
Fit for Purpose:
Certified Quality
Sterile
Sterile and PCR Ready
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Filtered Sterility Fit for Purpose
10371034 200 μL Non-filtered Sterile Sterile
10371033 50 μL Filtered Sterile Sterile and PCR Ready
10371032 50 μL Non-filtered Sterile Sterile
21403444 50 μL Non-filtered Non-sterile Certified Quality
21402305 100 μL Non-filtered Non-sterile Certified Quality
21402300 150 μL Filtered Sterile Sterile and PCR Ready
21402302 200 μL Non-filtered Non-sterile Certified Quality
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 10371034 Supplier Thermo Scientific™ Supplier No. MB923261

Increase pipetting performance with these automation tips, which undergo a rigorous 15-point quality control program to ensure accuracy and consistency.

Increase pipetting performance with Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Caliper/Zymark RapidPlate, Caliper Presto, Allegro, Zephyr and SciClone.

Features:

Specifications

Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Format Rack
Tip Style Automation
For Use With (Equipment) Zymark/Caliper Liquid Handling Workstations
Product Line Caliper Zymark
Fit for Purpose Sterile