missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations
GREENER_CHOICE

Catalog No. 10371033
Change view
Click to view available options
Volume (Metric):
50 μL
100 μL
150 μL
200 μL
Filtered:
Filtered
Non-filtered
Sterility:
Non-sterile
Sterile
Fit for Purpose:
Certified Quality
Sterile
Sterile and PCR Ready
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Filtered Sterility Fit for Purpose
10371033 50 μL Filtered Sterile Sterile and PCR Ready
10371032 50 μL Non-filtered Sterile Sterile
21403444 50 μL Non-filtered Non-sterile Certified Quality
21402305 100 μL Non-filtered Non-sterile Certified Quality
21402300 150 μL Filtered Sterile Sterile and PCR Ready
10371034 200 μL Non-filtered Sterile Sterile
21402302 200 μL Non-filtered Non-sterile Certified Quality
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 10371033 Supplier Thermo Scientific™ Supplier No. 920021

Increase pipetting performance with these automation tips, which undergo a rigorous 15-point quality control program to ensure accuracy and consistency.

Increase pipetting performance with Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Caliper/Zymark RapidPlate, Caliper Presto, Allegro, Zephyr and SciClone.

  • Available in non-sterile, sterile and with Thermo Scientific™ ART™ filter technology
  • Lot-tested during production on actual workstation for which they are created
  • Low percentage coefficient of variation (%CV)

Specifications

Volume (Metric) 50 μL
Filtered Filtered
Sterility Sterile
Color Clear
Format Rack
Tip Style Automation
For Use With (Equipment) Zymark/Caliper Liquid Handling Workstations
Product Line Caliper Zymark
Fit for Purpose Sterile and PCR Ready