Learn More
Aushon Chemiluminescent Mouse Angiogenesis Array Kits DFS Item
For simultaneous measurement of combinations of angiogenesis targets
Supplier:  Aushon Biosystems Inc 200101FA
 View more versions of this product
Catalog No. 200101FA
Only null left
Explore available promotions
Description
Description
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles
Specifications
Specifications
For simultaneous measurement of combinations of angiogenesis targets
Angiogenesis Array Kits
1 Plex
Product Suggestions
Product Suggestions