Learn More
Aushon Chemiluminescent Mouse Angiogenesis Array Kits
Catalog No. p-4480444
Encompass
Change view
Click to view available options
Quantity:
1 Plex
2 Plex
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
200041FA 1 Plex
200081FA 1 Plex
200101FA 1 Plex
201801FA 2 Plex
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 200041FA Supplier Aushon Biosystems Inc Supplier No. 200041FA
Only null left
For simultaneous measurement of combinations of angiogenesis targets
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles
Specifications
For Use With (Application) For simultaneous measurement of combinations of angiogenesis targets
Product Type Angiogenesis Array Kits
Quantity 1 Plex