Learn More
Aushon Chemiluminescent Mouse Angiogenesis Array Kits
For simultaneous measurement of combinations of angiogenesis targets
Supplier: Aushon Biosystems Inc 200081FA
Description
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles
Specifications
| For simultaneous measurement of combinations of angiogenesis targets | |
| Angiogenesis Array Kits | |
| 1 Plex |