missing translation for 'onlineSavingsMsg'
Learn More

Promega AttoPhos™ AP Fluorescent Substrate

Catalog No. prs1011
Encompass
Change view
Click to view available options
Quantity:
36 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRS1011 36 mg
PRS1012 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRS1011 Supplier Promega Supplier No. S1011

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

AttoPhos Substrate

A highly sensitive, flluorescent alkaline phosphatase substrate.

  • Produces a low fluorescence signal until enzymatically acted upon, yielding detection of AP to 0.1 attomole
  • Provides low background
  • Offers linear kinetics over five orders of magnitude of AP concentration
  • Has an unusually large Stokes' shift (≈120nm), which leads to higher detection sensitivity

Specifications

Form Powder
Quantity 36 mg
Substrate Type AttoPhos Substrate BBT
Storage Requirements 4°C
Substrate Quantity 36mg