missing translation for 'onlineSavingsMsg'
Learn More

Promega AttoPhos™ AP Fluorescent Substrate

A highly sensitive, flluorescent alkaline phosphatase substrate.

$210.81 - $370.24

Specifications

Form Powder
Substrate Type AttoPhos Substrate BBT
Storage Requirements 4°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRS1012
Encompass
View Documents Promotion Details
Promega
S1012
100 mg
Each for $370.24
Only null left
 
PRS1011
Encompass
View Documents Promotion Details
Promega
S1011
36 mg
Each for $210.81
Only null left
 
Description

Description

  • Produces a low fluorescence signal until enzymatically acted upon, yielding detection of AP to 0.1 attomole
  • Provides low background
  • Offers linear kinetics over five orders of magnitude of AP concentration
  • Has an unusually large Stokes' shift (≈120nm), which leads to higher detection sensitivity

Specifications

Specifications

Powder
AttoPhos Substrate BBT
4°C
Videos
SDS
Documents

Documents