missing translation for 'onlineSavingsMsg'
Learn More

Atomoxetine hydrochloride, Thermo Scientific™

Catalog No. AC467680010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467680010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC467680010 Supplier Thermo Scientific Chemicals Supplier No. 467680010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 82248-59-7
Molecular Formula C17H22ClNO
Molecular Weight (g/mol) 291.82
MDL Number 06410992
InChI Key LUCXVPAZUDVVBT-UNTBIKODSA-N
IUPAC Name hydrogen methyl[(3R)-3-(2-methylphenoxy)-3-phenylpropyl]amine chloride
SMILES [H+].[Cl-].CNCC[C@@H](OC1=CC=CC=C1C)C1=CC=CC=C1

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Physical Form Powder
Color White to Beige
Melting Point 166°C to 168°C
Solubility Soluble in methanol
Quantity 1 g
Formula Weight 291.82
Chemical Name or Material Atomoxetine hydrochloride
ShelfLife 5 years
Recommended Storage Refrigerator +4°C