missing translation for 'onlineSavingsMsg'
Learn More

Atipamezole Hydrochloride 98.0+%, TCI America™

Catalog No. p-7118168
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mg
25 mg
Catalog No. Quantity
A295625MG 25 mg
A2956100MG 100 mg
2 options

Catalog No. A295625MG

Supplier: TCI America A295625MG

Only null left

Chemical Identifiers

CAS 104075-48-1
Molecular Formula C14H17ClN2
Molecular Weight (g/mol) 248.75
MDL Number MFCD06407819
InChI Key PCCVCJAQMHDWJY-UHFFFAOYSA-N
Synonym atipamezole hydrochloride, atipamezole hcl, 1h-imidazole, 4-2-ethyl-2,3-dihydro-1h-inden-2-yl-, monohydrochloride, 4-2-ethyl-2,3-dihydro-1h-inden-2-yl-1h-imidazole hydrochloride, 5-2-ethyl-2,3-dihydro-1h-inden-2-yl-1h-imidazole hydrochloride, pubchem21910, mpv1248 hydrochloride, mpv 1248 hydrochloride, mpv-1248 hydrochloride
PubChem CID 13649426
IUPAC Name 5-(2-ethyl-2,3-dihydro-1H-inden-2-yl)-1H-imidazole hydrochloride
SMILES Cl.CCC1(CC2=CC=CC=C2C1)C1=CN=CN1

Specifications

Melting Point 220°C
Color White-Yellow
Quantity 25 mg
Formula Weight 248.75
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Atipamezole Hydrochloride
RTECSNumber NI6139000
TSCA No