CAS RN 104075-48-1
IUPAC Name:
5-(2-ethyl-2,3-dihydro-1H-inden-2-yl)-1H-imidazole hydrochloride
Synonyms:
atipamezole hydrochloride
Molecular Weight (g/mol):
248.75
Molecular Formula:
C14H17ClN2
InChi Key:
PCCVCJAQMHDWJY-UHFFFAOYSA-N
SMILES:
Cl.CCC1(CC2=CC=CC=C2C1)C1=CN=CN1
1
–
1
of
1
results
1
Atipamezole Hydrochloride 98.0+%, TCI America™
CAS: 104075-48-1 Molecular Formula: C14H17ClN2 Molecular Weight (g/mol): 248.75 MDL Number: MFCD06407819 InChI Key: PCCVCJAQMHDWJY-UHFFFAOYSA-N Synonym: atipamezole hydrochloride,atipamezole hcl,1h-imidazole, 4-2-ethyl-2,3-dihydro-1h-inden-2-yl-, monohydrochloride,4-2-ethyl-2,3-dihydro-1h-inden-2-yl-1h-imidazole hydrochloride,5-2-ethyl-2,3-dihydro-1h-inden-2-yl-1h-imidazole hydrochloride,pubchem21910,mpv1248 hydrochloride,mpv 1248 hydrochloride,mpv-1248 hydrochloride PubChem CID: 13649426 IUPAC Name: 5-(2-ethyl-2,3-dihydro-1H-inden-2-yl)-1H-imidazole hydrochloride SMILES: Cl.CCC1(CC2=CC=CC=C2C1)C1=CN=CN1