missing translation for 'onlineSavingsMsg'
Learn More

Atipamezole Hydrochloride 98.0+%, TCI America™

Catalog No. A295625MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A295625MG 25 mg
A2956100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A295625MG Supplier TCI America Supplier No. A295625MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 104075-48-1
Molecular Formula C14H17ClN2
Molecular Weight (g/mol) 248.75
MDL Number MFCD06407819
InChI Key PCCVCJAQMHDWJY-UHFFFAOYSA-N
Synonym atipamezole hydrochloride, atipamezole hcl, 1h-imidazole, 4-2-ethyl-2,3-dihydro-1h-inden-2-yl-, monohydrochloride, 4-2-ethyl-2,3-dihydro-1h-inden-2-yl-1h-imidazole hydrochloride, 5-2-ethyl-2,3-dihydro-1h-inden-2-yl-1h-imidazole hydrochloride, pubchem21910, mpv1248 hydrochloride, mpv 1248 hydrochloride, mpv-1248 hydrochloride
PubChem CID 13649426
IUPAC Name 5-(2-ethyl-2,3-dihydro-1H-inden-2-yl)-1H-imidazole hydrochloride
SMILES Cl.CCC1(CC2=CC=CC=C2C1)C1=CN=CN1

Specifications

Melting Point 220°C
Color White-Yellow
Quantity 25 mg
Formula Weight 248.75
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Atipamezole Hydrochloride
RTECSNumber NI6139000
TSCA No