missing translation for 'onlineSavingsMsg'
Learn More

Atazanavir, Thermo Scientific Chemicals

Catalog No. AC460981000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460981000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC460981000 Supplier Thermo Scientific Chemicals Supplier No. 460981000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 198904-31-3
Molecular Formula C38H52N6O7
Synonym zrivada

Specifications

Quantity 100 mg
Melting Point 207.0°C to 209.0°C
Formula Weight 704.86
Purity ≥97.5% (HPLC)
Chemical Name or Material Atazanavir

RUO – Research Use Only