missing translation for 'onlineSavingsMsg'
Learn More

Astragaloside IV, Thermo Scientific™

Catalog No. AC468862500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468862500 250 mg
AC468860010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC468862500 Supplier Thermo Scientific Chemicals Supplier No. 468862500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 84687-43-4
Molecular Formula C41 H68 O14

Specifications

Quantity 250 mg
Formula Weight 784.97
Physical Form Powder
Color White to Almost-white
Purity ≥97.5%
Chemical Name or Material Astragaloside IV
Recommended Storage Normal conditions