missing translation for 'onlineSavingsMsg'
Learn More

Astragaloside IV, Thermo Scientific™

Catalog No. AC468860010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468860010 1 g
AC468862500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC468860010 Supplier Thermo Scientific Chemicals Supplier No. 468860010

Chemical Identifiers

CAS 84687-43-4
Molecular Formula C41 H68 O14

Specifications

Quantity 1 g
Formula Weight 784.97
MDL Number MFCD16036240,MFCD16036240
Physical Form Powder
Color White to Almost-white
Purity ≥97.5%
InChI Key QMNWISYXSJWHRY-CSXKERSZSA-N
SMILES CC(C)(O)[C@@H]1CC[C@@](C)(O1)[C@H]1[C@H](O)C[C@@]2(C)C3C[C@@H](O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)C4[C@]5(CC35CC[C@]12C)CCC(O[C@@H]1OC[C@@H](O)[C@H](O)[C@H]1O)C4(C)C
IUPAC Name (2R,3R,4S,5S,6R)-2-{[(3R,9R,12S,14R,15R,16R)-14-hydroxy-15-[(2R,5S)-5-(2-hydroxypropan-2-yl)-2-methyloxolan-2-yl]-7,7,12,16-tetramethyl-6-{[(2S,3R,4S,5R)-3,4,5-trihydroxyoxan-2-yl]oxy}pentacyclo[9.7.0.0¹,³.0³,⁸.0¹²,¹⁶]octadecan-9-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol) 784.98
Chemical Name or Material Astragaloside IV
Show More Show Less
Recommended Storage Normal conditions