missing translation for 'onlineSavingsMsg'
Learn More

ASTM™ D6584 Tricaprin Solution, 8000 μg/mL in Pyridine, MilliporeSigma™ Supelco™

Catalog No. 111017769
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017769 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017769 Supplier Merck Emd Millipore Supplier No. CRM44897

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Petroleum

Specifications

CAS 621-71-6
Product Line ASTM
Flash Point 17.0°C - closed cup
Concentration 8,000 μg/mL in pyridine
Synonym Biodiesel impurity
Type Certified Reference Material
Quantity 5 mL
Grade TraceCERT™ Analytical Standard

ASTM is a registered trademark of American Society for Testing and Materials