missing translation for 'onlineSavingsMsg'
Learn More

ASTM™ D6584 Monoolein Solution, 5,000 μg/mL in Pyridine, MilliporeSigma™ Supelco™

Catalog No. 111017906
Change view
Click to view available options
Quantity:
3 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017906 3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017906 Supplier Merck Emd Millipore Supplier No. CRM44893

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Petroleum

Specifications

Packaging Ampule
Product Line ASTM
CAS 111-03-5
Flash Point 17.0°C - Closed Cup
Synonym Monoolein solution
Type Certified Reference Material
Quantity 3 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 5,000 μg/mL in pyridine

ASTM is a registered trademark of American Society for Testing and Materials