missing translation for 'onlineSavingsMsg'
Learn More

ASTM™ D5442 C12-C60 Qualitative Retention Time Mix, 6.25% (w/w) Each Component, Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111004680
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111004680 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111004680 Supplier Merck Emd Millipore Supplier No. 500623

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ANALYSIS OF PETROLEUM Waxes BY GC - Qualitative and quantitative mixes of n-paraffins are used for determining column resolution, retention times, and response factors.

Specifications

Concentration 6.25% (w/w) each component
Quantity 500 mg
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) Docosane; Dodecane; Dotriacontane; Eicosane; Hexacosane; Hexadecane; Hexatriacontane; Octacosane; Octadecane; Pentacontane; Tetracontane; Tetracosane; Tetradecane; Tetratetracontane; Triacontane

ASTM is a registered trademark of American Society for Testing and Materials