missing translation for 'onlineSavingsMsg'
Learn More

ASTM™ D4815 Qualitative Peak ID Mix Analytical Standard, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022543
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022543 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022543 Supplier Merck Emd Millipore Supplier No. 47213

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ASTM(R) D4815 Qualitative Peak ID Mix is for use in determination of oxygenates (Ethers and Alcohols) in gasoline by GC.

Specifications

Packaging Ampule
Flash Point 10°C
UN Number UN 1993 BE 3 / PGII
Quantity 1 mL
Grade Analytical Standard

ASTM is a registered trademark of American Society for Testing and Materials