missing translation for 'onlineSavingsMsg'
Learn More

ASTM™ D2887 Reference Gas Oil Analytical Standard, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022618
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022618 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022618 Supplier Merck Emd Millipore Supplier No. 506419

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This sample is a petroleum fraction with a boiling range from 250° to 850°F, evaluated in round-robin studies by the ASTM. Use this sample and the supplied ASTM boiling range consensus values to evaluate system performance.

Specifications

Packaging Ampule
Flash Point -38°C
Physical Form Neat
UN Number UN1203 - Class 3 - PG 2 - Gasoline
Synonym RGO
Quantity 1 mL
Grade Analytical Standard

ASTM is a registered trademark of American Society for Testing and Materials