missing translation for 'onlineSavingsMsg'
Learn More

ASTM™ D2887 Quantitative Calibration Solution Analytical Standard, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022603
Change view
Click to view available options
Quantity:
6 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022603 6 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022603 Supplier Merck Emd Millipore Supplier No. 500658

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Packaging Ampule
Flash Point -30°C
Concentration 0.5 wt. % (except where noted)
UN Number UN1131 - Class 3 - PG 1 - Carbon disulfide
Quantity 6 x 1 mL
Grade Analytical Standard

ASTM is a registered trademark of American Society for Testing and Materials