missing translation for 'onlineSavingsMsg'
Learn More

Medicom™ AssureTouch™ Ultra Tough Nitrile Powder-Free Medical Examination Gloves with Textured Fingertips

AssureTouch™ Ultra Tough Nitrile Powder-Free Medical Examination Gloves with Textured Fingertips. Industrial quality nitrile ensures high chemical and tear resistance without compromising comfort or fit. Fully textured to ensure a secure grip.

$143.13 - $143.13

Specifications

Chlorinated No
Accelerator Free No
Applications Healthcare, Industrial, Laboratory
AQL 1.5
Color Black
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17101093
View Documents
Medicom™
9950B
Small
Case of 1000 Each for $143.13
Only null left
 
17101094
View Documents
Medicom™
9950C
Medium
Case of 1000 Each for $143.13
Only null left
 
17101095
View Documents
Medicom™
9950D
Large
Case of 1000 Each for $143.13
Only null left
 
17101096
View Documents
Medicom™
9950E
X-Large
Case of 1000 Each for $143.13
Only null left
 
17101097
View Documents
Medicom™
9950F
2X-Large
Case of 1000 Each for $143.13
Only null left
 
Description

Description

  • Made from industrial quality nitrile to provide strong tear resistance without compromising comfort or fit.
  • Textured fingertips ensure a secure grip, even when wet.
Specifications

Specifications

No
Healthcare, Industrial, Laboratory
Black
Fingertip Textured
Powder Free
Yes
No
1.5
Single-use
Yes
Non-sterile
Beaded
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'