Learn More
MilliporeSigma™ Supelco™ Assembled Screw Cap with Hole
The septa and closure was designed to provide a positive physical lock to assure that preassembled components arrive intact.
Supplier:  MilliporeSigma™ Supelco™ 27261U
 View more versions of this product
Catalog No. 111016080
Only null left
Description
Description
  • The septa and closure was designed to provide a positive physical lock to assure that preassembled components arrive intact.
  • Screw caps are available for use with sealing septa.
  • Polypropylene caps have a maximum temperature of 135° C.
  • The PTFE/silicone septum can be used with most organics (until pierced) and is not recommended for chlorosilanes.
Specifications
Chemical Identifiers
PTFE, Silicone, Polypropylene
Specifications
Black
100
2 mL Vial with 4.6 mm Opening
PTFE, Silicone, Polypropylene
8-425
Product Suggestions
Product Suggestions