missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Assembled Screw Cap with Hole

Catalog No. p-7204516
Change view
Click to view available options
For Use With (Equipment):
15 mL Vial
2 mL Screw Thead Vial
2 mL Vial (Large Opening)
2 mL Vial (Standard Opening)
2 mL Vial with 4.6 mm Opening
20, 40, or 60 mL Vial
4 mL Vial
7 mL Vial
Color:
Black
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Color
111016080 2 mL Vial with 4.6 mm Opening Black
111016286 20, 40, or 60 mL Vial Black
111016289 2 mL Vial (Standard Opening) Black
111010439 15 mL Vial Black
111010711 7 mL Vial Black
111010834 4 mL Vial Black
111011455 2 mL Screw Thead Vial Black
111012072 2 mL Vial (Large Opening) Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 111016080 Supplier MilliporeSigma™ Supelco™ Supplier No. 27261U
Only null left

The septa and closure was designed to provide a positive physical lock to assure that preassembled components arrive intact.

  • The septa and closure was designed to provide a positive physical lock to assure that preassembled components arrive intact.
  • Screw caps are available for use with sealing septa.
  • Polypropylene caps have a maximum temperature of 135° C.
  • The PTFE/silicone septum can be used with most organics (until pierced) and is not recommended for chlorosilanes.

Specifications

Color Black
Material PTFE, Silicone, Polypropylene
No. per Pack 100
Closure Size 8-425
For Use With (Equipment) 2 mL Vial with 4.6 mm Opening