Learn More
Thermo Scientific Chemicals Ascorbate oxidase, from Cucurbita sp.
Catalog No. AAJ657824I
Change view
Click to view available options
Quantity:
1 kU
Catalog No. Quantity
AAJ657824I 1 kU
1 options
Catalog No. AAJ657824I Supplier Thermo Scientific Chemicals Supplier No. J657824I
Ascorbate oxidase, from Cucurbita sp., may be used to study oxidative stress and heat stress response and tolerance. Also used in ascorbic acid assays to study the heat stress response of Arabidopsis and useful for enzymatic determination of ascorbic acid and for eliminating the interference of ascorbic acid in clinical analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity 1 kU
Form Lyophilized
RUO – Research Use Only