missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Ascentis™ Si Supelguard™ Guard Cartridge Kit, 5 μm Particle Size

Catalog No. 111009604
Change view
Click to view available options
Diameter: 4 mm; Length: 2 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
111009604 Diameter: 4 mm; Length: 2 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111009604 Supplier MilliporeSigma™ Supelco™ Supplier No. 581519U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Ascentis™ family of columns is the fourth generation of HPLC column technology from Supelco scientists. Ascentis™ columns are bonded on high purity, 100 Angstrom silica including 3, 5, and 10 micron particle size. Columns are designed for small molecule applications and are scalable from micro columns (1.0 mm I.D.) to preparative dimensions (50 mm I.D.). The family includes C18, C8, Phenyl, Si and embedded polar group phase, RP-Amide.

Specifications

Quantity 1 Kit
Particle Size 5 μm
Length (Metric) 2 cm
Type Fused-Core HPLC Guard Cartridge Kit
Diameter (Metric) Inner 4 mm
Includes 1 cartridge, a stand-alone holder, a piece of tubing, and 2 nuts and ferrules
Phase Normal Phase and HILIC
For Use With (Application) HPLC
Product Line Ascentis Supelguard

Ascentis and Supelguard are trademarks of Sigma-Aldrich Co. LLC