Learn More
MilliporeSigma Ascentis™ Express PCS-Phenyl-Hexyl HPLC Column
Ascentis™ Express PCS-Phenyl-Hexyl columns are specifically engineered to provide efficient separation of basic, acidic, or neutral compounds under low ionic strength (formic acid) mobile phase conditions.
$1518.76 - $1993.38
Specifications
| Particle Size | 2.7 μm |
|---|---|
| Pore Size | 90 Å |
| Phase | Positively Charged Surface (PCS) |
| Material | Stainless Steel |
| Type | HPLC Column |
| Catalog Number | Mfr. No. | Diameter (Metric) | Length (Metric) | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Diameter (Metric) | Length (Metric) | Price | Quantity | ||||
|
01-000-896
|
MilliporeSigma
50702U |
4.6 mm | 150 mm |
Each for $1,871.33
|
|
||||
|
01-000-897
|
MilliporeSigma
50700U |
4.6 mm | 100 mm |
Each for $1,695.05
|
|
||||
|
01-000-898
|
MilliporeSigma
50697U |
4.6 mm | 50 mm |
Each for $1,518.76
|
|
||||
|
01-000-899
|
MilliporeSigma
50696U |
3.0 mm | 150 mm |
Each for $1,871.33
|
|
||||
|
01-000-900
|
MilliporeSigma
50694U |
3.0 mm | 100 mm |
Each for $1,695.05
|
|
||||
|
01-000-901
|
MilliporeSigma
50693U |
3.0 mm | 50 mm |
Each for $1,518.76
|
|
||||
|
01-000-902
|
MilliporeSigma
50692U |
2.1 mm | 150 mm |
Each for $1,871.33
|
|
||||
|
01-000-903
|
MilliporeSigma
50691U |
2.1 mm | 100 mm |
Each for $1,695.05
|
|
||||
|
01-000-904
|
MilliporeSigma
50690U |
2.1 mm | 50 mm |
Each for $1,518.76
|
|
||||
|
01-000-905
|
MilliporeSigma
50689U |
1.5 mm | 150 mm |
Each for $1,993.38
|
|
||||
|
01-000-906
|
MilliporeSigma
50688U |
1.5 mm | 100 mm |
Each for $1,830.66
|
|
||||
|
01-000-907
|
MilliporeSigma
50686U |
1.5 mm | 50 mm |
Each for $1,654.36
|
|
||||
Description
These columns contain advanced Fused-Core™ particles with pores strategically sized to 90 Å for small molecule analyses. This attribute makes these columns an excellent choice for pharmaceutical method development.
- The unique PCS-C18 chemistry enables exceptional peak shape and enhanced loading capacity of basic compounds as compared to traditional C18 chemistries.
- This column also provides orthogonal selectivity to other small molecule chemistries.
Specifications
| 2.7 μm | |
| Positively Charged Surface (PCS) | |
| HPLC Column |
| 90 Å | |
| Stainless Steel | |
| HPLC |