missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma Ascentis™ Express HPLC Column, 2.7 μm

Catalog No. M50515U
Change view
Click to view available options
Diameter (Metric):
2.1 mm
3.0 mm
4.6 mm
Length (Metric):
5 cm
10 cm
15 cm
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Length (Metric)
M50515U 4.6 mm 15 cm
M50502U 2.1 mm 5 cm
M50503U 2.1 mm 10 cm
M50504U 2.1 mm 15 cm
M50505U 3.0 mm 5 cm
M50506U 3.0 mm 10 cm
M50510U 3.0 mm 15 cm
M50513U 4.6 mm 5 cm
M50514U 4.6 mm 10 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. M50515U Supplier MilliporeSigma Supplier No. 50515U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

HPLC column for chromatography applications.

  • This product is a Design for Sustainability (DfS) developed Greener Alternative product, which belongs to one of the four categories of greener alternatives.
  • Has a phosphorylcholinezwitterionic stationary phase covalently attached to Fused-Core™ particle.
  • The permanent and hydrophiliczwitterion functionality makes the column suitable for Hydrophilic Interaction Liquid Chromatography (HILIC).
  • Weak electrostatic interactions between charged analytes and the neutral zwitterionic stationary phase results in a unique selectivity, and especially suitable for analytes that are poorly retained on reversed phase columns.
  • Can be used as a tool to change the selectivity or to improve peak resolution for polar and hydrophilic compounds such as carbohydrates, metabolites, acids and bases, organic and inorganic ions, metal complexes, amino acids, peptides, and protein digests.

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 15 cm
Particle Size 2.7 μm
Pore Size 160 Å
Phase ZIC-cHILIC Phase
Material Stainless Steel
Technique HPLC, LC/MS