missing translation for 'onlineSavingsMsg'
Learn More

Ascentis™ Express 160 Å Phenyl-Hexyl, 2.7 μm HPLC Column, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022324
Change view
Click to view available options
Length (Metric):
3 cm
10 cm
15 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
111022324 3 cm
111022322 10 cm
111022323 15 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111022324 Supplier Merck Emd Millipore Supplier No. 584617U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Target analytes include electron-poor molecules, aromatic or unsaturated compounds (ketones, nitriles, alkenes), as well as peptides and polypeptides < 20 kDa.

  • High-speed, high-performance liquid chromatography column based on a 160Å Fused-Core™ particle design
  • The particle provides a thin porous shell of high-purity silica surrounding a solid silica core.
  • This particle design exhibits very high column efficiency for high molecular weight solutes (up to 20 kDa) due to the shallow diffusion paths in the 0.5 μm thick porous shell and the small overall particle size of 2.7 μm.
  • The endcapped, phenyl-hexyl bonded phase provides a stable, reversed phase packing with a pore structure and pore size that is optimized for reversed-phase HPLC separations of peptides and polypeptides, using typical acidic mobile phases favored for proteomic applications.

Specifications

pH 2 to 9
Particle Size 2.7 μm
Length (Metric) 3 cm
Pore Size 160 Å pore size
Surface Area 90 m2/g
Diameter (Metric) Inner 4.6 mm
For Use With (Equipment) L11 Per USP
Stationary Phase Phenyl-Hexyl (dimethylphenyl-hexylsilane) bonding
Endcapped Endcapped
Matrix Fused-Core™ particle platform

Ascentis is a registered trademark of Sigma-Aldrich Co. LLC, Fused-Core is a registered trademark of Advanced Materials Technology, Inc.