missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ASB 14780
Description
ASB 14780 is a potent and selective cytosolic phospholipase A2 alpha (cPLA2α) inhibitor (IC50 = 20 nM). Selective for cPLA2α over secreted PLA2α (sPLA2α; exhibits no inhibition at 10 μM). Inhibits cPLA2α-dependent inflammatory responses in vitro in guinea pig and human whole-blood assays. Improves diet-induced liver injury and CCl4-induced hepatic fibrosis in vivo. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 3-(3-Phenethyl-1-(4-phenoxyphenyl)-1 H-indol-5-yl)propanoic acid tris salt |
| CAS | 1069046-00-9 |
| Quantity | 5 mg |
| Target | Phospholipase Inhibitors |
| Molecular Formula | C31H27NO3.C4H11NO3 |
| Purity | 0.98 |