missing translation for 'onlineSavingsMsg'
Learn More

Arsenic Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC PLAS22X

 View more versions of this product

Catalog No. PLAS2-2X


Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
As1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Arsenic Standard
7440-38-2,7697-37-2,7732-18-5
As
AA, ICP
Certified Reference Material
[As]
arsenic
500 mL
CHEBI:27563
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
MFCD00085309
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
RQNWIZPPADIBDY-UHFFFAOYSA-N
2% HNO3
74.92
5359596
Assurance

Chemical Identifiers

7440-38-2
74.92
RQNWIZPPADIBDY-UHFFFAOYSA-N
CHEBI:27563
[As]
As
MFCD00085309
5359596
arsenic
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Causes severe skin burns and eye damage.

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only