missing translation for 'onlineSavingsMsg'
Learn More

Arsenic, AAS standard solution, Specpure™ As 1000μg/mL

$79.27 - $153.75

Chemical Identifiers

MDL Number MFCD00085309
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA33559AE
View Documents
Thermo Scientific Chemicals
033559AE
100 mL
Each for $79.27
Only null left
 
AA33559AP
View Documents
Thermo Scientific Chemicals
033559AP
500 mL
Each for $153.75
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

MFCD00085309

Specifications

Liquid
MFCD00085309
100 mL
Arsenic AAS standard solution, As 1000μg/mL
As in 5% HNO3
UN3264
Specpure
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314
Causes severe skin burns and eye damage.

P201-P202-P234-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335-H350

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.

EINECSNumber : 231-714-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only