Learn More
Arsenic, AAS standard solution, Specpure™ As 1000μg/mL
Supplier: Thermo Scientific Chemicals 033559AE
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Arsenic AAS standard solution | |
| Liquid | |
| MFCD00085309 | |
| 100 mL |
| As 1000μg/mL | |
| As in 5% HNO3 | |
| UN3264 | |
| Specpure |
Chemical Identifiers
| MFCD00085309 |
Safety and Handling
GHS H Statement
H314
Causes severe skin burns and eye damage.
P201-P202-P234-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335-H350
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber : 231-714-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only