missing translation for 'onlineSavingsMsg'
Learn More

Aroclor 1254, High Level, SPEX CertiPrep™

Catalog No. pcb1254h
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PCB1254H 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PCB1254H Supplier SPEX CertiPrep Group LLC Supplier No. SPX PCB 1254H

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Aroclor 125411097-69-11000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 11097-69-1 , 110-54-3
Molecular Formula C12H5Cl5
Molecular Weight (g/mol) 326.422
InChI Key AUGNBQPSMWGAJE-UHFFFAOYSA-N
PubChem CID 40470
IUPAC Name 1,2,3-trichloro-4-(2,3-dichlorophenyl)benzene
SMILES C1=CC(=C(C(=C1)Cl)Cl)C2=C(C(=C(C=C2)Cl)Cl)Cl

Specifications

Packaging 2mL amber prescored glass ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
CAS Max % 99.9, 0.1
Methods For US EPA Methods 505, 508, 625, 8082, 8270C
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Hexane
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL
Chemical Name or Material Aroclor 1254 (High Level)
Hazard Category Highly flammable liquid and vapor. Causes serious eye irritation. May cause drowsiness or dizziness.
DOTInformation Nonhazardous
Recommended Storage Ambient

Product intended for laboratory use only