missing translation for 'onlineSavingsMsg'
Learn More

Aroclor 1254, SPEX CertiPrep™

Catalog No. PCB1254
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PCB1254 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PCB1254 Supplier SPEX CertiPrep Group LLC Supplier No. PCB1254

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Aroclor 125411097-69-1200μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 11097-69-1 , 110-54-3
Molecular Formula C12H5Cl5
Molecular Weight (g/mol) 326.422
InChI Key AUGNBQPSMWGAJE-UHFFFAOYSA-N
PubChem CID 40470
IUPAC Name 1,2,3-trichloro-4-(2,3-dichlorophenyl)benzene
SMILES C1=CC(=C(C(=C1)Cl)Cl)C2=C(C(=C(C=C2)Cl)Cl)Cl

Specifications

Packaging 2mL amber prescored glass ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
CAS Min % 0.02, 99.98
CAS Max % 0.02, 99.98
Methods For US EPA Methods 505, 508, 608, 625, 8082, 8270C
Technique AA, ICP
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Hexane
Quantity 1 mL
Chemical Name or Material Aroclor 1254
Show More Show Less
DOTInformation Nonhazardous
Recommended Storage Ambient

Product intended for laboratory use only