missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals (-)-Arctiin

Catalog No. AC466050010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466050010 1 g
AC466052500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466050010 Supplier Thermo Scientific Chemicals Supplier No. 466050010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20362-31-6
Molecular Formula C27H34O11

Specifications

Quantity 1 g
Melting Point 109.0°C to 112.0°C
Formula Weight 534.56
InChI Key XOJVHLIYNSOZOO-RVDKUWPISA-N
SMILES COC1=CC=C(C[C@H]2COC(=O)[C@@H]2CC2=CC=C(OC3OC(CO)C(O)C(O)C3O)C(OC)=C2)C=C1OC
IUPAC Name (3R,4R)-4-[(3,4-dimethoxyphenyl)methyl]-3-[(3-methoxy-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}phenyl)methyl]oxolan-2-one
Molecular Weight (g/mol) 534.56
Chemical Name or Material (-)-Arctiin

RUO – Research Use Only