missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-Arctigenin 95.0+%, TCI America™
Supplier: TCI America A311750MG
Specifications
| (-)-Arctigenin | |
| 100°C | |
| C21H24O6 | |
| 50 mg | |
| NQWVSMVXKMHKTF-JKSUJKDBSA-N | |
| (3R,4R)-4-[(3,4-dimethoxyphenyl)methyl]-3-[(4-hydroxy-3-methoxyphenyl)methyl]oxolan-2-one | |
| 64981 | |
| 372.42 | |
| Crystalline Powder |
| 7770-78-7 | |
| White-Yellow | |
| MFCD00870597 | |
| arctigenin, --arctigenin, unii-u76mr9vs6m, u76mr9vs6m, 2 3h-furanone, 4-3,4-dimethoxyphenyl methyl dihydro-3-4-hydroxy-3-methoxyphenyl methyl-, 3r-trans, 2 3h-furanone, 4-3,4-dimethoxyphenyl methyl dihydro-3-4-hydroxy-3-methoxyphenyl methyl-, 3r,4r, 3r,4r-4-3,4-dimethoxyphenyl methyl-3-4-hydroxy-3-methoxyphenyl methyl oxolan-2-one, 3r,4r-4-3,4-dimethoxyphenyl methyl dihydro-3-4-hydroxy-3-methoxyphenyl methyl-2 3h-furanone, +/--arctigenin, arctigenin, +/- | |
| COC1=C(C=C(C=C1)CC2COC(=O)C2CC3=CC(=C(C=C3)O)OC)OC | |
| 372.417 | |
| CHEBI:79 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 7770-78-7 | |
| 372.417 | |
| NQWVSMVXKMHKTF-JKSUJKDBSA-N | |
| 64981 | |
| (3R,4R)-4-[(3,4-dimethoxyphenyl)methyl]-3-[(4-hydroxy-3-methoxyphenyl)methyl]oxolan-2-one |
| C21H24O6 | |
| MFCD00870597 | |
| arctigenin, --arctigenin, unii-u76mr9vs6m, u76mr9vs6m, 2 3h-furanone, 4-3,4-dimethoxyphenyl methyl dihydro-3-4-hydroxy-3-methoxyphenyl methyl-, 3r-trans, 2 3h-furanone, 4-3,4-dimethoxyphenyl methyl dihydro-3-4-hydroxy-3-methoxyphenyl methyl-, 3r,4r, 3r,4r-4-3,4-dimethoxyphenyl methyl-3-4-hydroxy-3-methoxyphenyl methyl oxolan-2-one, 3r,4r-4-3,4-dimethoxyphenyl methyl dihydro-3-4-hydroxy-3-methoxyphenyl methyl-2 3h-furanone, +/--arctigenin, arctigenin, +/- | |
| CHEBI:79 | |
| COC1=C(C=C(C=C1)CC2COC(=O)C2CC3=CC(=C(C=C3)O)OC)OC |
Safety and Handling
RTECSNumber : LY9247000
TSCA : No