missing translation for 'onlineSavingsMsg'
Learn More

(-)-Arctigenin 95.0+%, TCI America™

Catalog No. p-7118225
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A311710MG 10 mg
A311750MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A311710MG Supplier TCI America Supplier No. A311710MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7770-78-7
Molecular Formula C21H24O6
Molecular Weight (g/mol) 372.417
MDL Number MFCD00870597
InChI Key NQWVSMVXKMHKTF-JKSUJKDBSA-N
Synonym arctigenin, --arctigenin, unii-u76mr9vs6m, u76mr9vs6m, 2 3h-furanone, 4-3,4-dimethoxyphenyl methyl dihydro-3-4-hydroxy-3-methoxyphenyl methyl-, 3r-trans, 2 3h-furanone, 4-3,4-dimethoxyphenyl methyl dihydro-3-4-hydroxy-3-methoxyphenyl methyl-, 3r,4r, 3r,4r-4-3,4-dimethoxyphenyl methyl-3-4-hydroxy-3-methoxyphenyl methyl oxolan-2-one, 3r,4r-4-3,4-dimethoxyphenyl methyl dihydro-3-4-hydroxy-3-methoxyphenyl methyl-2 3h-furanone, +/--arctigenin, arctigenin, +/-
PubChem CID 64981
ChEBI CHEBI:79
IUPAC Name (3R,4R)-4-[(3,4-dimethoxyphenyl)methyl]-3-[(4-hydroxy-3-methoxyphenyl)methyl]oxolan-2-one
SMILES COC1=C(C=C(C=C1)CC2COC(=O)C2CC3=CC(=C(C=C3)O)OC)OC

Specifications

Melting Point 100°C
Color White-Yellow
Quantity 10 mg
Formula Weight 372.42
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (-)-Arctigenin
RTECSNumber LY9247000
TSCA No