Learn More
National Safety Apparel™ Enespro™ ArcGuard™ 25 cal Coat/Bib Kit, No Gloves
Arc flash kit with double layer Ultrasoft™
$2644.50 - $2877.24
Specifications
Ankle Style Open
Arc Flash Rating 25 cal/cm²
Certifications/Compliance CAT 3, NFPA 70E, ASTM F1506, CSA Z462, OSHA 1910.269
Closure Type FR Hook and Loop
Color Khaki
View More Specs
Products 8
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
170011414
View Documents
National Safety Apparel™
KIT3SC25NGSM
Small
Each for $2,644.50
Only null left
170011415
View Documents
National Safety Apparel™
KIT3SC25NGMD
Medium
Each for $2,644.50
Only null left
170011416
View Documents
National Safety Apparel™
KIT3SC25NGLG
Large
Each for $2,644.50
Only null left
170011417
View Documents
National Safety Apparel™
KIT3SC25NGXL
X-Large
Each for $2,644.50
Only null left
170011418
View Documents
National Safety Apparel™
KIT3SC25NG2X
2X-Large
Each for $2,722.07
Only null left
170011419
View Documents
National Safety Apparel™
KIT3SC25NG3X
3X-Large
Each for $2,722.07
Only null left
170011420
View Documents
National Safety Apparel™
KIT3SC25NG4X
4X-Large
Each for $2,799.65
Only null left
170011421
View Documents
National Safety Apparel™
KIT3SC25NG5X
5X-Large
Each for $2,877.24
Only null left
Description
Description
  • Faceshield Bag
  • Faceshield with hood and hard hat
  • Large Gear Bag
  • Safety Glasses
  • UltraSoft™ Bib Overall
  • UltraSoft™ Short Coat
Specifications
Specifications
Open
CAT 3, NFPA 70E, ASTM F1506, CSA Z462, OSHA 1910.269
Khaki
Arc Flash Kit
Adjustable
Unisex
25 cal/cm²
FR Hook and Loop
Cotton, Nylon
Stitched
Utilities, Manufacturing, Electrical Contractors