Learn More
National Safety Apparel™ Enespro™ ArcGuard™ 25 cal Coat/Bib Kit, No Gloves
Arc flash kit with double layer Ultrasoft™
Supplier: National Safety Apparel™ KIT3SC25NG3X
Description
- Faceshield Bag
- Faceshield with hood and hard hat
- Large Gear Bag
- Safety Glasses
- UltraSoft™ Bib Overall
- UltraSoft™ Short Coat
Specifications
| 25 cal/cm² | |
| CAT 3, NFPA 70E, ASTM F1506, CSA Z462, OSHA 1910.269 | |
| Utilities, Manufacturing, Electrical Contractors | |
| Cotton, Nylon | |
| Unisex | |
| Stitched |
| Khaki | |
| Arc Flash Kit | |
| Open | |
| 3X-Large | |
| FR Hook and Loop | |
| Adjustable |