missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 25 cal Coat/Bib Kit, No Gloves

Catalog No. 170011414
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
170011414 Small
170011415 Medium
170011416 Large
170011417 X-Large
170011418 2X-Large
170011419 3X-Large
170011420 4X-Large
170011421 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 170011414 Supplier National Safety Apparel™ Supplier No. KIT3SC25NGSM

Arc flash kit with double layer Ultrasoft™

  • Faceshield Bag
  • Faceshield with hood and hard hat
  • Large Gear Bag
  • Safety Glasses
  • UltraSoft™ Bib Overall
  • UltraSoft™ Short Coat

Specifications

For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Gender Unisex
Certifications/Compliance CAT 3, NFPA 70E, ASTM F1506, CSA Z462, OSHA 1910.269
Product Type Arc Flash Kit
Arc Flash Rating 25 cal/cm²
Suspender Adjustable
Material Cotton, Nylon
Size Small
Closure Type FR Hook and Loop
Seam Type Stitched
Ankle Style Open
Color Khaki
Show More Show Less