missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ AquaPhenol™
Description
Phenol is used as a reagent in chemical analysis. It is used in the manufacture of colorless or light-colored artificial resins, many medical and industrial organic compounds and dyes. It is commonly used in tissue preservation and DNA/RNA extraction with chloroform.
Specifications
Specifications
| Content And Storage | Store at +4°C. Store under nitrogen. Protect from light. |
| Formulation | Water saturated Phenol |
| pH | 4.5 - 5 (Lit.) |
| Research Category | Molecular Biology |
| Quantity | 250 mL |
| Form | Solution |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.