missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Karl Fischer Aqualine™ Solvent CM, for Karl Fischer Titration By Volumetry, Fisher Chemical™
$2,388.23 - $2,388.23
Specifications
| Chemical Name or Material | Aqualine™ Solvent CM |
|---|---|
| Name Note | Karl Fischer Reagent |
| CAS | 288-32-4 , 67-66-3 , 7446-09-5 |
| Physical Form | Liquid |
| MDL Number | 81734 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AL-2110212
|
Fisher Chemical
AL2110212 |
2.5 L |
N/A
|
|
|||||
Description
- Solvent for samples with high hydrocarbon content
- Used in conjunction with Aqualine™titrant for two component volumetric Karl Fischer analysis