missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Karl Fischer Aqualine™ Complete 1, for Karl Fischer Titration By Volumetry, Fisher Chemical™
Single-component reagent; Pyridine free
$229.71 - $229.71
Specifications
| Chemical Name or Material | Aqualine™ Complete 1 |
|---|---|
| Name Note | Karl Fischer Reagent |
| Color | Colorless |
| CAS | 7446-09-5 , 288-32-4 , 111-90-0 , 7553-56-2 |
| Physical Form | Liquid |
Description
- Single-component volumetric reagent for high water content analysis
- Water equivalent: 1.0mg H2O/mL
- Unlimited water capacity
- Perform titer determination before use
- Pyridine free