Learn More
Apramycin sulfate, 96%
Potent inhibitor of protein synthesis in bacteria | CAS: 65710-07-8 | C21H43N5O15S | 637.66 g/mol
Supplier: Thermo Scientific Chemicals J6387406
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
ApplicationsPotent inhibitor of protein synthesis in bacteria.Apramycin sulfate is used as a potent inhibitor of protein synthesis in bacteria. It acts as an antibacterial agent. It is a useful biochemical for antibiotic and antibacterial research to study antibiotic resistance through protein synthesis inhibition by blocking translocation. It is also essential for clinic in vitro tests known as antimicrobial susceptibility tests to determine their efficacy against certain bacterial species.
Solubility
Soluble in water.
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.Incompatible with strong oxidizing agents.
Specifications
| Apramycin Sulfate | |
| Powder | |
| 168°C | |
| 5 g | |
| C21H43N5O15S | |
| 823°C | |
| Soluble in water. | |
| OS(O)(=O)=O.CN[C@H]1[C@@H](O)[C@H]2O[C@H](O[C@@H]3[C@@H](N)C[C@@H](N)[C@H](O)[C@H]3O)[C@H](N)C[C@@H]2O[C@@H]1O[C@H]1O[C@H](CO)[C@@H](N)[C@H](O)[C@H]1O | |
| 637.66 | |
| 637.65 |
| Apramycin sulfate | |
| 65710-07-8 | |
| White to Yellow | |
| 451.6°C (845°F) | |
| MFCD06200257 | |
| 14,752 | |
| WGLYHYWDYPSNPF-RQFIXDHTSA-N | |
| (2R,3R,4S,5S,6S)-2-{[(2R,3S,4R,4aR,6S,7R,8aS)-7-amino-6-{[(1R,2R,3S,4R,6S)-4,6-diamino-2,3-dihydroxycyclohexyl]oxy}-4-hydroxy-3-(methylamino)-octahydropyrano[3,2-b]pyran-2-yl]oxy}-5-amino-6-(hydroxymethyl)oxane-3,4-diol; sulfuric acid | |
| 3081544 | |
| 96% |
Chemical Identifiers
| 65710-07-8 | |
| 637.66 | |
| WGLYHYWDYPSNPF-RQFIXDHTSA-N | |
| (2R,3R,4S,5S,6S)-2-{[(2R,3S,4R,4aR,6S,7R,8aS)-7-amino-6-{[(1R,2R,3S,4R,6S)-4,6-diamino-2,3-dihydroxycyclohexyl]oxy}-4-hydroxy-3-(methylamino)-octahydropyrano[3,2-b]pyran-2-yl]oxy}-5-amino-6-(hydroxymethyl)oxane-3,4-diol; sulfuric acid |
| C21H43N5O15S | |
| MFCD06200257 | |
| 3081544 | |
| OS(O)(=O)=O.CN[C@H]1[C@@H](O)[C@H]2O[C@H](O[C@@H]3[C@@H](N)C[C@@H](N)[C@H](O)[C@H]3O)[C@H](N)C[C@@H]2O[C@@H]1O[C@H]1O[C@H](CO)[C@@H](N)[C@H](O)[C@H]1O |
Safety and Handling
GHS H Statement
H360-H315-H319
May damage fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
P201-P202-P264b-P280i-P281-P302+P352-P305+P351+P338-P308+P313-P332+P313-P362-P501c
H315-H319-H360
EINECSNumber : 265-890-7
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only