missing translation for 'onlineSavingsMsg'
Learn More

Apramycin sulfate, 96%

Potent inhibitor of protein synthesis in bacteria | CAS: 65710-07-8 | C21H43N5O15S | 637.66 g/mol

$193.25 - $938.37

Chemical Identifiers

CAS 65710-07-8
Molecular Formula C21H43N5O15S
Molecular Weight (g/mol) 637.66
MDL Number MFCD06200257
InChI Key WGLYHYWDYPSNPF-RQFIXDHTSA-N
PubChem CID 3081544
IUPAC Name (2R,3R,4S,5S,6S)-2-{[(2R,3S,4R,4aR,6S,7R,8aS)-7-amino-6-{[(1R,2R,3S,4R,6S)-4,6-diamino-2,3-dihydroxycyclohexyl]oxy}-4-hydroxy-3-(methylamino)-octahydropyrano[3,2-b]pyran-2-yl]oxy}-5-amino-6-(hydroxymethyl)oxane-3,4-diol; sulfuric acid
SMILES OS(O)(=O)=O.CN[C@H]1[C@@H](O)[C@H]2O[C@H](O[C@@H]3[C@@H](N)C[C@@H](N)[C@H](O)[C@H]3O)[C@H](N)C[C@@H]2O[C@@H]1O[C@H]1O[C@H](CO)[C@@H](N)[C@H](O)[C@H]1O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ6387403
View Documents Promotion Details
N/A 1 g
Each for $193.25
Only null left
 
AAJ6387406
View Documents Promotion Details
N/A 5 g
Each for $938.37
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Potent inhibitor of protein synthesis in bacteria.Apramycin sulfate is used as a potent inhibitor of protein synthesis in bacteria. It acts as an antibacterial agent. It is a useful biochemical for antibiotic and antibacterial research to study antibiotic resistance through protein synthesis inhibition by blocking translocation. It is also essential for clinic in vitro tests known as antimicrobial susceptibility tests to determine their efficacy against certain bacterial species.

Solubility
Soluble in water.

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

65710-07-8
637.66
WGLYHYWDYPSNPF-RQFIXDHTSA-N
(2R,3R,4S,5S,6S)-2-{[(2R,3S,4R,4aR,6S,7R,8aS)-7-amino-6-{[(1R,2R,3S,4R,6S)-4,6-diamino-2,3-dihydroxycyclohexyl]oxy}-4-hydroxy-3-(methylamino)-octahydropyrano[3,2-b]pyran-2-yl]oxy}-5-amino-6-(hydroxymethyl)oxane-3,4-diol; sulfuric acid
C21H43N5O15S
MFCD06200257
3081544
OS(O)(=O)=O.CN[C@H]1[C@@H](O)[C@H]2O[C@H](O[C@@H]3[C@@H](N)C[C@@H](N)[C@H](O)[C@H]3O)[C@H](N)C[C@@H]2O[C@@H]1O[C@H]1O[C@H](CO)[C@@H](N)[C@H](O)[C@H]1O

Specifications

Apramycin Sulfate
65710-07-8
White to Yellow
C21H43N5O15S
823°C
Soluble in water.
OS(O)(=O)=O.CN[C@H]1[C@@H](O)[C@H]2O[C@H](O[C@@H]3[C@@H](N)C[C@@H](N)[C@H](O)[C@H]3O)[C@H](N)C[C@@H]2O[C@@H]1O[C@H]1O[C@H](CO)[C@@H](N)[C@H](O)[C@H]1O
637.66
637.65
Apramycin sulfate
Powder
168°C
451.6°C (845°F)
MFCD06200257
14,752
WGLYHYWDYPSNPF-RQFIXDHTSA-N
(2R,3R,4S,5S,6S)-2-{[(2R,3S,4R,4aR,6S,7R,8aS)-7-amino-6-{[(1R,2R,3S,4R,6S)-4,6-diamino-2,3-dihydroxycyclohexyl]oxy}-4-hydroxy-3-(methylamino)-octahydropyrano[3,2-b]pyran-2-yl]oxy}-5-amino-6-(hydroxymethyl)oxane-3,4-diol; sulfuric acid
3081544
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360-H315-H319
May damage fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.

P201-P202-P264b-P280i-P281-P302+P352-P305+P351+P338-P308+P313-P332+P313-P362-P501c

H315-H319-H360

EINECSNumber : 265-890-7

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only