missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ True Allele™ PCR Premix

Catalog No. 403061
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
403061 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 403061 Supplier Applied Biosystems™ Supplier No. 403061
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

10 tubes, each containing 1.8mL of True Allele™ PCR Premix, enough to perform 2000 PCR amplifications of 15μL each.

Applied Biosystems True Allele PCR Premix is a PCR master mix that contains AmpliTaq Gold DNA Polymerase, dNTPs, MgCl2, and buffer.

Applied Biosystems True Allele PCR Premix is a PCR master mix that contains AmpliTaq Gold DNA Polymerase, dNTPs, MgCl2, and buffer. It is intended for use with the ABI PRISM Linkage Mapping Set. True Allele PCR Premix has been optimized for strong, specific PCR amplification of microsatellite markers.

Accessory products

ABI PRISM Linkage Mapping Set

Notes

See user's manual or package insert for limited label license and trademark information.

Order Info

Shipping condition: Dry Ice

Specifications

Concentration 5 U/μL
Content And Storage 10 tubes of True Allele PCR Premix, 1.8 mL each

Store at -20°C.
Format Tube
Polymerase AmpliTaq DNA Polymerase
Reaction Speed Standard
Exonuclease Activity 5' - 3'
Product Type PCR Premix
Quantity 1 kit
Shipping Condition Dry Ice
For Use With (Application) Hot-start PCR
Fidelity (vs. Taq) 1X
Hot Start Built-In Hot Start
No. of Reactions 2000 Reactions
Reaction Format Separate Components
Size (Final Product) 5 kb or less
Starting Material DNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.