missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Universal PCR Master Mix, no AmpErase™ UNG
TaqMan™ Universal PCR Master Mix, No AmpErase™ UNG is the ideal reagent solution when you need a Master Mix for multiple 5' nuclease DNA applications.
Supplier: Applied Biosystems™ 4326614
Description
- Contains AmpliTaq Gold™ DNA polymerase to provide a better yield and a more robust 5' nuclease assay than AmpliTaq™ DNA polymerase
- Includes a passive internal reference based on proprietary ROX™ dye, for superb precision on Applied Biosystems™ real-time PCR instruments
- Buffer enhancements guarantee performance and reliability in all applications, even with G/C-rich sequences
- Contains dUTP in the dNTP mix to protect against carry-over contamination, when used with AmpErase™ UNG
- Rapid assay development guidelines are provided to minimize optimization time
Order Info
Guaranteed minimum shelf life is 60 days. Shipping Conditions: Wet Ice.
Chemical Identifiers
| Primer-probe | |
| qPCR | |
| Standard | |
| ROX (Pre-mixed) | |
| Universal PCR Master Mix | |
| Gene Expression, Genotyping, miRNA analysis |
| High | |
| AmpliTaq Gold DNA Polymerase | |
| 7000 System, 7300 System, 7500 System, 7700 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, GeneAmp 9600, StepOne™, Standard Mode, StepOnePlus™, Standard Mode | |
| TaqMan | |
| Wet Ice | |
| DNA (Genomic), cDNA |
Specifications
| Supplied at 2X concentration. The mix contains AmpliTaq Gold™ DNA Polymerase, dNTPs with dUTP, Passive Reference 1 and optimized buffer components. This pack contains 1 X 50 mL tubes, sufficient for 2000 reactions at the 50 μL total volume. Store at 2-8°C Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). |
|
| High | |
| AmpliTaq Gold DNA Polymerase | |
| 7000 System, 7300 System, 7500 System, 7700 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, GeneAmp 9600, StepOne™, Standard Mode, StepOnePlus™, Standard Mode | |
| TaqMan | |
| 1 x 50 mL 2,000 reactions | |
| 2000 Reactions | |
| 2000 Reactions | |
| 50 mL |
| Primer-probe | |
| qPCR | |
| Standard | |
| ROX (Pre-mixed) | |
| Universal PCR Master Mix | |
| Wet Ice | |
| Gene Expression, Genotyping, miRNA analysis | |
| DNA (Genomic), cDNA |
For Research Use Only. Not for use in diagnostic procedures.